Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cystatin B Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Cystatin B/Stefin B |
---|---|
Clone | 227807 |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14091SP
![]() |
R&D Systems
MAB14091SP |
25 μg |
Each for $200.50
|
|
|||||
MAB14091
![]() |
R&D Systems
MAB14091 |
500 μg |
Each for $643.50
|
|
|||||
Description
Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Mouse samples. It is validated for Immunohistochemistry.Specifications
Cystatin B/Stefin B | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
CSTB | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
227807 | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q62426 | |
1476 | |
E. coli-derived recombinant mouse Cystatin B Met2-Phe98 Accession # Q62426 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Cystatin B in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin B is observed and no cross-reactivity with rhCystatin A, C, D, E/M, F, S, SA, rhFetuin A, rhFetuin B, rhHPRG, or rhKininogen is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title