Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cystatin B Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1409
This item is not returnable.
View return policy
Description
Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin B/Stefin B | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1476 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunoprecipitation | |
227818 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q62426 | |
CSTB | |
E. coli-derived recombinant mouse Cystatin B Met2-Phe98 Accession # Q62426 | |
500 μg | |
Primary | |
Detects mouse Cystatin B in direct ELISAs and Western blots. In Western blots, 20-25% cross-reactivity with recombinant human (rh) Cystatin B is observed and no cross-reactivity with rhCystatin D, S, SA, SN, recombinant mouse (rm) Cystatin A, E/M, rmFetuin A, rhFetuin B, rhHPRG, or rhKininogen is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction