Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Cystatin/Stefin Homolog 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1516SP
This item is not returnable.
View return policy
Description
Cystatin/Stefin Homolog 2 Monoclonal specifically detects Cystatin/Stefin Homolog 2 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin/Stefin Homolog 2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
NP_084009 | |
2010005H15Rik | |
E. coli-derived recombinant mouse Cystatin/Stefin Homolog 2 Met2-Phe97 Accession # NP_084009 | |
25 μg | |
Primary | |
Detects mouse Cystatin/Stefin Homolog 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, S, SA, SN, recombinant mouse Cystain B, C, E/M, rhFetuin A, rhFetuin B, rhHPRG, or rhKininostatin is observed. | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
237305 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
010005H15Rik, Csta2, Cystatin H2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
76770 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction