Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DAN Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7552SP
This item is not returnable.
View return policy
Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
DAN/NBL1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4681 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, ELISA | |
123224 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q61477 | |
NBL1 | |
Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477 | |
25 μg | |
Primary | |
Detects mouse DAN in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) DAN, rhCOCO, recombinant mouse (rm) BMP-4, rmGremlin, or recombinant chicken (rch) Caronte is observed. In Western blots, approximately 10% cross-reactivity with rhDAN, rhCOCO, and rchCaronte is observed and no cross reactivity with rmGremlin is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction