Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DAN Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | DAN/NBL1 |
---|---|
Clone | 123202 |
Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Blockade of Receptor-ligand Interaction |
Classification | Monoclonal |
Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
DAN/NBL1 | |
Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
NBL1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
123202 | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Rat | |
Mouse | |
Q61477.2 | |
4681 | |
Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse DAN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) DAN, recombinant chicken Caronte, rhCerberus, or recombinant mouse Gremlin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title