Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DAN Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | DAN/NBL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
DAN/NBL1 Polyclonal specifically detects DAN/NBL1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
DAN/NBL1 | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Mouse | |
DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
NBL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q499K5 | |
4681 | |
Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q499K5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse DAN in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title