Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DC-SIGN/CD209 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB83451RS100UG
This item is not returnable.
View return policy
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry.Specifications
DC-SIGN/CD209 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Purified mouse DC-SIGN/CD209 extracellular domain Accession # FJ168685 | |
100 μg | |
30835 | |
Mouse | |
Supernatant |
Flow Cytometry | |
MMD3R | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
FJ168685 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse DC-SIGN/CD209 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction