Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DC-SIGN/CD209 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB8345SP
This item is not returnable.
View return policy
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
DC-SIGN/CD209 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
30835 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
902404 | |
Flow Cytometry 0.25 ug/mL, CyTOF-ready | |
Q91ZX1 | |
Cd209a | |
NS0 mouse myeloma cell line transfected with mouse DC-SIGN/CD209 Met1-Lys238 Accession # Q91ZX1 | |
25 μg | |
Primary | |
Detects mouse DC-SIGN/CD209 in direct ELISAs. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction