Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DC-SIGN/CD209 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $502.50
Specifications
Antigen | DC-SIGN/CD209 |
---|---|
Clone | MMD3R |
Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
Applications | CyTOF, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB83451R-SP
![]() |
R&D Systems
MAB83451RSP |
25 μg |
Each for $214.00
|
|
|||||
AB83451R100
![]() |
R&D Systems
MAB83451R100 |
100 μg |
Each for $502.50
|
|
|||||
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
DC-SIGN/CD209 | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
FJ168685 | |
30835 | |
IgG2c | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
MMD3R | |
CyTOF, Flow Cytometry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Purified mouse DC-SIGN/CD209 extracellular domain Accession # FJ168685 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects mouse DC-SIGN/CD209 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title