Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DCAR/CLEC4B Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2757G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2757G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2757G100 Supplier R&D Systems Supplier No. FAB2757G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

DCAR/CLEC4B Monoclonal specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen DCAR/CLEC4B
Applications Flow Cytometry
Classification Monoclonal
Clone 349214
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAR13070
Gene Alias Aplra2, CLEC4B, CLECSF3, LSLCL, P47, SCGF
Gene Symbols Clec4b1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B Gln42-Leu209 Accession # AAR13070
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Test Specificity Detects mouse DCAR/CLEC4B in direct ELISAs.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.