Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DCAR/CLEC4B Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB2757N100UG
This item is not returnable.
View return policy
Description
DCAR/CLEC4B Monoclonal specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Flow Cytometry.Specifications
DCAR/CLEC4B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAR13070 | |
Clec4b1 | |
Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B Gln42-Leu209 Accession # AAR13070 | |
RUO | |
6320 | |
Mouse | |
IgG2a |
Flow Cytometry | |
349214 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Aplra2, CLEC4B, CLECSF3, LSLCL, P47, SCGF | |
Rat | |
100 μg | |
Primary | |
Detects mouse DCAR/CLEC4B in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction