Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DCAR/CLEC4B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2757
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2757 100 μg
AF2757SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2757 Supplier R&D Systems Supplier No. AF2757
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DCAR/CLEC4B Polyclonal specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen DCAR/CLEC4B
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q7TS58
Gene Alias Aplra2, CLEC4B, CLECSF3, LSLCL, P47, SCGF
Gene Symbols Clec4b1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B Gln42-Leu209 Accession # Q7TS58
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Test Specificity Detects mouse DCAR/CLEC4B in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse DCIR is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.