Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DCIR/CLEC4A PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB26171P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB26171P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB26171P Supplier R&D Systems Supplier No. FAB26171P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen DCIR/CLEC4A
Applications Flow Cytometry
Classification Monoclonal
Clone 320507
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9QZ15
Gene Alias CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR
Gene Symbols Clec4a2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DCIR/CLEC4A isoform 1 Gln70-Leu238 Accession # Q9QZ15
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50856
Test Specificity Detects mouse DCIR/CLEC4A in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.