Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DcTRAILR1/TNFRSF23 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2378SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2378SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2378SP Supplier R&D Systems Supplier No. MAB2378SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

DcTRAILR1/TNFRSF23 Monoclonal specifically detects DcTRAILR1/TNFRSF23 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen DcTRAILR1/TNFRSF23
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 270012
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9ER63
Gene Symbols Tnfrsf23
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23 Met30-Ser156 Accession # Q9ER63
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Detects mouse DcTRAIL R1/TNFRSF23 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant mouse (rm) 4-1BB Ligand, recombinant human (rh) BAFF R, rhDcTRAIL R3, rmFas, rmOPG, and rhTRAIL R4 is observed and no cross-reactivity with rmCD27, rmCD30, rmDR3, rhDR6, rmEDAR, recombinant feline Fas, rhFas, recombinant rat Fas, rmGITR, rmHVEM, rmLymphotoxin beta R, rmNGF R, rmOX40, rmTNF RI, rmTNF RII, rhTRAIL R1, or rhTROY is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.