Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DEC-205/CD205 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5975
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5975 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5975 Supplier R&D Systems Supplier No. AF5975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DEC-205/CD205 Polyclonal specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen DEC-205/CD205
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q60767
Gene Symbols LY75
Host Species Goat
Immunogen E. coli-derived recombinant mouse DEC-205/CD205 Cys216-Pro503 Accession # Q60767
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects mouse DEC-205/CD205 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human DEC-205 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.