Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Decorin Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $585.50
Specifications
Antigen | Decorin |
---|---|
Clone | 161026 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Decorin Monoclonal specifically detects Decorin in Mouse samples. It is validated for Western Blot.Specifications
Decorin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P28654 | |
1634 | |
Mouse myeloma cell line NS0-derived recombinant mouse Decorin Gly17-Lys354 Accession # P28654 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Decorin in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human Decorin. |
161026 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B | |
DCN | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title