Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Dectin-1/CLEC7A APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB17561A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB17561A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB17561A Supplier R&D Systems Supplier No. FAB17561A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Dectin-1/CLEC7A
Applications Flow Cytometry
Classification Monoclonal
Clone 218820
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q6QLQ4
Gene Alias Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Gene Symbols CLEC7A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Dectin-1/CLEC7A Phe69-Leu244 Accession # Q6QLQ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64581
Test Specificity Detects mouse Dectin-1/CLEC7A in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Dectin-1 is observed and no cross-reactivity with recombinant mouse Dectin-2 or rhDLEC is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.