Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Dectin-2/CLEC6A alpha Isoform Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1525SP
This item is not returnable.
View return policy
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Western Blot.Specifications
Dectin-2/CLEC6A | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9JKF4 | |
Clec4n | |
Mouse myeloma cell line NS0-derived recombinant mouse Dectin-2/CLEC6A Phe43-Leu209 Accession # Q9JKF4 | |
25 μg | |
Primary | |
Detects mouse Dectin-2/CLEC6A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Dectin-1 or rhDLEC is observed. | |
Mouse | |
Purified |
Western Blot | |
217611 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
93978 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction