Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Dectin-2/CLEC6A alpha Isoform Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $561.00
Specifications
Antigen | Dectin-2/CLEC6A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Dectin-2/CLEC6A Polyclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Western Blot.Specifications
Dectin-2/CLEC6A | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
Clec4n | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9JKF4 | |
93978 | |
Mouse myeloma cell line NS0-derived recombinant mouse Dectin-2/CLEC6A Phe43-Leu209 Accession # Q9JKF4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Dectin-2/CLEC6A alpha Isoform in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human DLEC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title