Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Dectin-2/CLEC6A alpha Isoform APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB1525A
This item is not returnable.
View return policy
Description
Dectin-2/CLEC6A Polyclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Flow Cytometry.Specifications
Dectin-2/CLEC6A | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
Goat | |
Affinity Purified | |
RUO | |
93978 | |
Mouse | |
IgG |
Flow Cytometry | |
APC | |
Q9JKF4 | |
Clec4n | |
Mouse myeloma cell line NS0-derived recombinant mouse Dectin-2 alpha/CLEC6A Phe43-Leu209 Accession # Q9JKF4 | |
100 Tests | |
Primary | |
Detects mouse Dectin-2 alpha/CLEC6A alpha Isoform in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human DLEC is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction