Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Dectin-2/CLEC6A alpha Isoform Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1525
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1525 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1525 Supplier R&D Systems Supplier No. BAF1525
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Dectin-2/CLEC6A Polyclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Dectin-2/CLEC6A
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9JKF4
Gene Alias CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Gene Symbols Clec4n
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Dectin-2 alpha/CLEC6A Phe43-Leu209 Accession # Q9JKF4
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Test Specificity Detects mouse Dectin-2 alpha/CLEC6A alpha Isoform in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human DLEC is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.