Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Desert Hedgehog/Dhh C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF196
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF196 Supplier R&D Systems Supplier No. AF196
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Desert Hedgehog/Dhh Polyclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Desert Hedgehog/Dhh
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q61488
Gene Alias desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145
Gene Symbols DHH
Host Species Goat
Immunogen E. coli-derived recombinant mouse Desert Hedgehog/Dhh C-Terminus Cys199-Gly396 Accession # Q61488
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50846
Test Specificity Detects mouse Desert Hedgehog/Dhh C-Terminus in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Shh C-terminus peptide (aa 199-437) and rmIhh C-terminus peptide (aa 241-449) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.