Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB733
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB733 500 μg
MAB733SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB733 Supplier R&D Systems Supplier No. MAB733
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Desert Hedgehog/Dhh Monoclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Desert Hedgehog/Dhh
Applications Western Blot
Classification Monoclonal
Clone 126925
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61488
Gene Alias desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145
Gene Symbols DHH
Host Species Rat
Immunogen E. coli-derived recombinant mouse Desert Hedgehog/Dhh Cys23-Gly396 Accession # Q61488
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50846
Test Specificity Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In Western blots, no cross-reactivity with the N-terminal peptides of recombinant human Shh or recombinant mouse (rm) Shh is observed. Also, in direct ELISAs and Western blots, no cross-reactivity with the C-terminal peptides of rmDhh, rmIhh, or rmShh is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.