Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Desert Hedgehog/Dhh N-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody

$188.28 - $567.90

Specifications

Antigen Desert Hedgehog/Dhh
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AF733SP
GREENER_CHOICE
View Documents
R&D Systems
AF733SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
AF733
GREENER_CHOICE
View Documents
R&D Systems
AF733
100 μg
Each for $567.90
Only null left
Add to Cart
 
Description

Description

Desert Hedgehog/Dhh Polyclonal specifically detects Desert Hedgehog/Dhh in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Desert Hedgehog/Dhh
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Polyclonal
Goat
Mouse
desert hedgehog, desert hedgehog (Drosophila) homolog, desert hedgehog homolog, desert hedgehog protein, DHH, HHG-3MGC35145
DHH
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot, Immunohistochemistry
Unconjugated
RUO
Q61488
50846
E. coli-derived recombinant mouse Desert Hedgehog/Dhh aa 23-198 Accession # Q61488
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse Desert Hedgehog/Dhh N-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse (rm) Shh N-terminal peptide (aa 25-198) is observed. Additionally, in direct ELISAs, no cross-reactivity with rmDhh C-terminal peptide (aa 199-396) and rmIhh C-terminal peptide (aa 241-449) is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.