Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Desmocollin-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7265SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7265SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7265SP Supplier R&D Systems Supplier No. MAB7265SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Desmocollin-3 Monoclonal specifically detects Desmocollin-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Desmocollin-3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 765129
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P55850
Gene Alias Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP
Gene Symbols DSC3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-3 Arg136-Leu695 Accession # P55850
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1825
Test Specificity Detects mouse Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human Desmocollin-3 is observed and no cross-reactivity with recombinant mouse (rm) Desmocollin-1 or rmDesmocollin-2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.