Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Dkk-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $549.50
Specifications
| Antigen | Dkk-3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Dkk-3 Polyclonal specifically detects Dkk-3 in Mouse samples. It is validated for Western Blot.Specifications
| Dkk-3 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk3, Dkk-3, hDkk-3, regulated in glioma, REIC, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| DKK3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9QUN9 | |
| 27122 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dkk-3 Met1-Ile349 Accession # Q9QUN9 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Dkk-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Dkk-3 is observed and less than 1% cross-reactivity with rhDkk-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title