Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DLL4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1389
This item is not returnable.
View return policy
Description
DLL4 Polyclonal specifically detects DLL4 in Mouse samples. It is validated for Western Blot.Specifications
DLL4 | |
Polyclonal | |
Biotin | |
BAB18580 | |
DLL4 | |
Mouse myeloma cell line NS0-derived recombinant mouse DLL4 Ser28-Pro525 Accession # BAB18580 | |
50 μg | |
Primary | |
Detects mouse DLL4 in Western blots. In this format, approximately 50% cross-reactivity with recombinant human (rh) DLL4 is observed and less than 5% cross-reactivity with rhDLL1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
Goat | |
Affinity Purified | |
RUO | |
54567 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction