Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DMBT1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5915G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5915G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5915G100 Supplier R&D Systems Supplier No. FAB5915G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

DMBT1/GP340 Monoclonal specifically detects DMBT1/GP340 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen DMBT1/GP340
Applications Flow Cytometry
Classification Monoclonal
Clone 548031
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q60997
Gene Alias CRP-ductin, deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, Ebnerin, FLJ61058, Glycoprotein 340, gp-340, GP340, GP340muclin, Hensin, MGC164738, Muclin, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein, Vomeroglandin
Gene Symbols DMBT1
Host Species Rat
Immunogen E. Coli-derived recombinant mouse DMBT1 Trp215-Gly420 Accession # Q60997
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1755
Test Specificity Detects mouse DMBT1 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CD163 and rhSRCRB4D is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.