Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DNAM-1/CD226 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4436 Supplier R&D Systems Supplier No. MAB4436
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen DNAM-1/CD226
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 838216
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8K4F0
Gene Alias CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Gene Symbols CD226
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DNAM-1/CD226 Met1-Pro254 Accession # Q8K4F0
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10666
Test Specificity Detects mouse DNAM-1/CD226 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DNAM-1/CD226 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.