Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DNAM-1/CD226 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF4436
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Polyclonal specifically detects DNAM-1/CD226 in Mouse samples. It is validated for Western Blot.Specifications
DNAM-1/CD226 | |
Polyclonal | |
Biotin | |
Q8K4F0 | |
CD226 | |
Mouse myeloma cell line NS0-derived recombinant mouse DNAM-1/CD226 Met1-Pro254 Accession # Q8K4F0 | |
50 μg | |
Primary | |
Detects mouse DNAM-1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human DNAM-1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
Sheep | |
Affinity Purified | |
RUO | |
10666 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction