Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DPPA5/ESG1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | DPPA5/ESG1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
DPPA5/ESG1 Polyclonal specifically detects DPPA5/ESG1 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
DPPA5/ESG1 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
developmental pluripotency associated 5, developmental pluripotency-associated 5 protein, embryonal stem cell specific gene 1, Embryonal stem cell-specific gene 1 protein, Esg1, ESG1ESG-1, hDPPA5 | |
Dppa5a | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q9CQS7 | |
340168 | |
E. coli-derived recombinant mouse DPPA5/ESG1 Met1-Glu118 Accession # Q9CQS7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse DPPA5/ESG1 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human DPPA5 is observed and approximately 10% cross-reactivity with recombinant mouse DPPA4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title