Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse DPPII/QPP/DPP7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3436
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3436 100 μg
AF3436SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3436 Supplier R&D Systems Supplier No. AF3436
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DPPII/QPP/DPP7 Polyclonal specifically detects DPPII/QPP/DPP7 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.

Specifications

Antigen DPPII/QPP/DPP7
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL
Gene Accession No. NP_114031
Gene Alias carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPP7, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase
Gene Symbols DPP7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DPPII/QPP/DPP7 Asp34-Arg506 Accession # NP_114031
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29952
Test Specificity Detects mouse DPPII/QPP/DPP7 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human DPPII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.