Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DPPIV/CD26 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB9541SP
This item is not returnable.
View return policy
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
DPPIV/CD26 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DPP4 | |
Mouse myeloma cell line NS0-derived recombinant mouse DPPIV/CD26 Extracellular domain | |
25 μg | |
Primary | |
Detects mouse DPPIV/CD26 in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
155202 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1803 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction