Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DPPIV/CD26 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF954
This item is not returnable.
View return policy
Description
DPPIV/CD26 Polyclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
DPPIV/CD26 | |
Polyclonal | |
Biotin | |
P28843 | |
DPP4 | |
Mouse myeloma cell line NS0-derived recombinant mouse DPPIV/CD26 Ser29-His760 Accession # P28843 | |
50 μg | |
Primary | |
Detects mouse DPPIV/CD26 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human DPPIV/CD26 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 | |
Goat | |
Affinity Purified | |
RUO | |
1803 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction