Learn More
Novus Biologicals™ Mouse dsDNA Autoantibody (IgG) ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum |
| Specificity | This assay has high sensitivity and excellent specificity for detection of mouse dsDNA autoantibody (IgG). No significant cross-reactivity or interference between mouse dsDNA autoantibody (IgG) and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Target | Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.