Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse DSPG3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2875SP
This item is not returnable.
View return policy
Description
DSPG3/EPYC Monoclonal specifically detects DSPG3/EPYC in Mouse samples. It is validated for Western Blot.Specifications
DSPG3/EPYC | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P70186 | |
EPYC | |
Mouse myeloma cell line NS0-derived recombinant mouse DSPG3 Ala20-Ile322 Accession # P70186 | |
25 μg | |
Primary | |
Detects mouse DSPG3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human DSPG3 is observed. | |
Mouse | |
Purified |
Western Blot | |
340908 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, EPYC, PG-Lb, PGLB, Proteoglycan-Lb, SLRR3B, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1833 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction