Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse E-Selectin/CD62E Biotinylated Antibody, R&D Systems™
Chicken Polyclonal Antibody
Supplier: R&D Systems BAF575
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
E-Selectin/CD62E | |
Polyclonal | |
Biotin | |
Q00690 | |
SELE | |
Mouse myeloma cell line NS0-derived recombinant mouse E-Selectin/CD62E Trp22-Pro557 Accession # Q00690 | |
50 μg | |
Primary | |
Detects mouse E-Selectin/CD62E in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human E-Selectin is observed and 5% cross-reactivity with recombinant mouse L-Selectin is observed. | |
Mouse | |
IgY |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
Chicken | |
Affinity Purified | |
RUO | |
6401 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction