Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse E-Selectin/CD62E Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems BAM5752
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
E-Selectin/CD62E | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
Q00690 | |
SELE | |
Mouse myeloma cell line NS0-derived recombinant mouse E-Selectin Trp22-Pro557 Accession # Q00690 | |
100 μg | |
Primary | |
Detects mouse E-Selectin/CD62E in ELISAs and Western blots. In Western blots, this antibody shows 20 - 50% cross-reactivity with recombinant human E-Selectin and recombinant mouse (rm) L-Selectin and no cross-reactivity with rmP-Selectin or recombinant rat E-Selectin. | |
Mouse | |
Purified |
Western Blot, ELISA | |
96409 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6401 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction