Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse EBI3 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC18341C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC18341C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC18341C Supplier R&D Systems Supplier No. IC18341C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EBI3 Monoclonal specifically detects EBI3 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Classification Monoclonal
Clone 355022
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O35228 (EBI3) & Q8K3I6 (p28)
Gene Alias cytokine receptor, EBI-3, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 beta, IL-27 subunit beta, IL27B, IL-27B, IL35 EBI3, IL-35 EBI3, interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer Tyr19-Pro228 (EBI3) & Phe29-Ser234 (p28) Accession # O35228 (EBI3) & Q8K3I6 (p28)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10148
Test Specificity Detects mouse IL-27/IL-35 EBI3 Subunit in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human IL-27/IL-35 EBI3 subunit.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.