Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EDAR Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
| Antigen | EDAR |
|---|---|
| Clone | 132106 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
EDAR Monoclonal specifically detects EDAR in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| EDAR | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| EDAR | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 132106 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9R187 | |
| 10913 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ile189 Accession # Q9R187 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse EDAR in ELISAs. In sandwich immunoassays, less than 10% cross-reactivity with recombinant human EDAR is observed and no cross-reactivity with recombinant mouse (rm) EDA, rmTROY, or rhXEDAR is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title