Learn More
Mouse EMMPRIN/CD147 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Specifications
Specifications
| Antigen | EMMPRIN/CD147 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1159A |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.1 ug/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P18572 |
| Gene Alias | 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.