Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EMMPRIN/CD147 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$198.10
Specifications
Antigen | EMMPRIN/CD147 |
---|---|
Clone | 1159A |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB7721R025
![]() |
R&D Systems
FAB7721R025UG |
25 μg |
Each of 1 for $198.10
|
|
|||||
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Flow Cytometry.Specifications
EMMPRIN/CD147 | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
IgG | |
Protein A or G purified from cell culture supernatant |
1159A | |
Monoclonal | |
Rabbit | |
Mouse | |
P18572 | |
682 | |
Primary | |
Detects mouse EMMPRIN/CD147 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title