Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EMMPRIN/CD147 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | EMMPRIN/CD147 |
---|---|
Clone | 1159A |
Dilution | Western Blot 1:1000 dilution, Immunohistochemistry 1:200-1:400 dilution |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EMMPRIN/CD147 | |
Western Blot 1:1000 dilution, Immunohistochemistry 1:200-1:400 dilution | |
Monoclonal | |
Purified | |
RUO | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
BSG | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Detects mouse EMMPRIN/CD147 in direct ELISAs. |
1159A | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse | |
P18572 | |
682 | |
NS0 mouse myeloma cell line transfected with mouse EMMPRIN/CD147 Ala22-Pro209 Accession # P18572 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title