Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse EMMPRIN/CD147 PE-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $374.00
Specifications
| Antigen | EMMPRIN/CD147 |
|---|---|
| Clone | 1159A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB7721P025
![]() |
R&D Systems
FAB7721P025UG |
25 μg |
Each for $227.50
|
|
|||||
FAB7721P100
![]() |
R&D Systems
FAB7721P100UG |
100 μg |
Each for $374.00
|
|
|||||
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Flow Cytometry.Specifications
| EMMPRIN/CD147 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Rabbit | |
| Mouse | |
| P18572 | |
| 682 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Detects mouse EMMPRIN/CD147 in direct ELISAs. |
| 1159A | |
| Flow Cytometry | |
| R-PE | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| NS0 mouse myeloma cell line transfected with mouse EMMPRIN/CD147 Ala22-Pro209 Accession # P18572 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title