Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Endoglin/CD105 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $1798.00
Specifications
| Antigen | Endoglin/CD105 |
|---|---|
| Clone | 209721 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13201SP
![]() |
R&D Systems
MAB13201SP |
25 μg |
Each for $200.50
|
|
|||||
MAB13201100
![]() |
R&D Systems
MAB13201100 |
100 μg |
Each for $491.00
|
|
|||||
MAB13201500
![]() |
R&D Systems
MAB13201500 |
500 μg |
Each for $1,798.00
|
|
|||||
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Endoglin/CD105 | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8K100 | |
| 2022 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Endoglin/CD105 Glu27-Gly581 Accession # Q8K100 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Endoglin/CD105 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human Endoglin. |
| 209721 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| ENG | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title