Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Endoglin/CD105 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1320
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1320 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1320 Supplier R&D Systems Supplier No. AF1320
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

Endoglin/CD105 Polyclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Endoglin/CD105
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. Q8K100
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1
Gene Symbols ENG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Endoglin/CD105 Glu27-Gly581 Accession # Q8K100
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Test Specificity Detects mouse Endoglin in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant rat Endoglin is observed and less than 5% cross-reactivity with recombinant human Endoglin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.