Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Endoglycan/PODXL2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3534
This item is not returnable.
View return policy
Description
Endoglycan/PODXL2 Polyclonal specifically detects Endoglycan/PODXL2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Endoglycan/PODXL2 | |
Polyclonal | |
Unconjugated | |
Q8CAE9 | |
PODXL2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Endoglycan/PODXL2 Gly30-Gly498 Accession # Q8CAE9 | |
100 μg | |
Primary | |
Detects mouse Endoglycan/PODXL2 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2, PODXL2 | |
Goat | |
Affinity Purified | |
RUO | |
50512 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction