Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Endomucin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF4666
This item is not returnable.
View return policy
Description
Endomucin/Mucin -14 Polyclonal specifically detects Endomucin/Mucin -14 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Endomucin/Mucin -14 | |
Polyclonal | |
Unconjugated | |
Q9R0H2 | |
EMCN | |
E. coli-derived recombinant mouse Endomucin Glu21-Glu90 Accession # Q9R0H2 | |
100 μg | |
Primary | |
Detects mouse Endomucin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Endomucin-2 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
EMCN, EMCN2, endomucin, MUC-14 | |
Goat | |
Affinity Purified | |
RUO | |
51705 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction