Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Endostatin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB570
This item is not returnable.
View return policy
Description
Endostatin Monoclonal specifically detects Endostatin in Mouse samples. It is validated for Western Blot.Specifications
| Endostatin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antiangiogenic agent, COL18A1, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP | |
| Rat | |
| Protein A or G purified from ascites | |
| RUO | |
| 80781 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 91318 | |
| Western Blot 1 ug/mL | |
| AAA19787 | |
| COL18A1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Endostatin His1105-Lys1288 Accession # AAA19787 | |
| 500 μg | |
| Primary | |
| Detects mouse Endostatin in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction