Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Endostatin ELISA Kit (Colorimetric)
SDP

Catalog No. NB293833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293833 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293833 Supplier Novus Biologicals™ Supplier No. NBP326055
Add to Cart
Add to Cart

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Mouse Endostatin in samples. No significant cross-reactivity or interference between Mouse Endostatin and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 80781
Gene Symbol COL18A1
Quantity 1 Kit
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.